Order Entry
Switzerland
ContactUsLinkComponent
(R)-5,6,7,8-tetrahydroquinolin-8-amine hydrochloride ≥98%
(R)-5,6,7,8-tetrahydroquinolin-8-amine hydrochloride ≥98%
Catalog # APOSOR510221-100MG
Supplier:  Apollo Scientific
CAS Number:  
(R)-5,6,7,8-tetrahydroquinolin-8-amine hydrochloride ≥98%
Catalog # APOSOR510221-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :184.666 g/mol
Storage Temperature:Ambient
CAS Number:1431726-92-9
Formula:C₉H₁₂N₂·ClH
MDL Number:MFCD31654063