Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromonaphthalen-1-ol
3-Bromonaphthalen-1-ol ≥97%
3-Bromonaphthalen-1-ol ≥97%
APOSOR510070-250MG
:
Apollo Scientific
:
3-Bromonaphthalen-1-ol ≥97%
APOSOR510070-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
223.066 g/mol
:
Refrigerator
:
90767-17-2
:
C₁₀H₇BrO
:
MFCD17012415