Order Entry
Switzerland
ContactUsLinkComponent
Methyl (2R)-4-oxopiperidine-2-carboxylate, N-BOC protected 95%
Methyl (2R)-4-oxopiperidine-2-carboxylate, N-BOC protected 95%
Katalognr. APOSOR50476-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl (2R)-4-oxopiperidine-2-carboxylate, N-BOC protected 95%
Katalognr. APOSOR50476-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :257.283 g/mol
Storage Temperature:Refrigerator
CAS Number:1799811-83-8
Formula:C₁₂H₁₉NO₅
MDL Number:MFCD12547038