
3-Amino-4-(4-methylphenyl)pyrrolidine, 3-BOC protected hydrochloride
Catalog # APOSOR48076-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:312.835 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:2367002-73-9
Formula:C₁₆H₂₄N₂O₂·ClH
MDL Number:MFCD32111679