Order Entry
Switzerland
ContactUsLinkComponent
(4-Chloro-2,5-dimethyl-phenyl)-(4-methylsulphonylphenyl)methanone 95%
(4-Chloro-2,5-dimethyl-phenyl)-(4-methylsulphonylphenyl)methanone 95%
Catalog # APOSOR480697-5G
Supplier:  Apollo Scientific
CAS Number:  
(4-Chloro-2,5-dimethyl-phenyl)-(4-methylsulphonylphenyl)methanone 95%
Catalog # APOSOR480697-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :322.806 g/mol
Storage Temperature:Ambient
CAS Number:1280729-47-6
Formula:C₁₆H₁₅ClO₃S
MDL Number:MFCD21365058