Order Entry
Switzerland
ContactUsLinkComponent
1-(1-(tert-butoxycarbonyl)-1H-indol-3-yl)cyclopropane-1-carboxylic acid
1-(1-(tert-butoxycarbonyl)-1H-indol-3-yl)cyclopropane-1-carboxylic acid
Catalog # APOSOR480136-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(1-(tert-butoxycarbonyl)-1H-indol-3-yl)cyclopropane-1-carboxylic acid
Catalog # APOSOR480136-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :301.337 g/mol
Storage Temperature:Ambient
Formula:C₁₇H₁₉NO₄
MDL Number:MFCD30496575