2-[4-(Cyclohexyloxy)phenyl]ethylamine
Catalog # APOSOR470904-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.323 g/mol
Storage Temperature:Ambient
CAS Number:1042579-53-2
Formula:C₁₄H₂₁NO
MDL Number:MFCD11167592