2-[4-(2-Cyclopropylethoxy)phenyl]ethylamine
Catalog # APOSOR470903-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.296 g/mol
Storage Temperature:Ambient
CAS Number:1481230-07-2
Formula:C₁₃H₁₉NO
MDL Number:MFCD21260218