Order Entry
Switzerland
ContactUsLinkComponent
(4-Bromo-2-chlorophenyl)(4-chlorophenyl)methanone
(4-Bromo-2-chlorophenyl)(4-chlorophenyl)methanone
Catalog # APOSOR470552-5G
Supplier:  Apollo Scientific
CAS Number:  
(4-Bromo-2-chlorophenyl)(4-chlorophenyl)methanone
Catalog # APOSOR470552-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :330.004 g/mol
Storage Temperature:Ambient
CAS Number:1157395-92-0
Formula:C₁₃H₇BrCl₂O
MDL Number:MFCD12065414