
2-(Bromomethyl)phenyl acetate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:704-65-4
:
Formula:C₉H₉BrO₂
MDL Number:MFCD20487652
:229.071 g/mol
Storage Temperature:Freezer
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together