
2,5-Dimethyl-4-nitro-2H-pyrazole-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3920-37-4
:
Formula:C₆H₇N₃O₄
MDL Number:MFCD00662656
:185.137 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together