Order Entry
Switzerland
ContactUsLinkComponent
4-Bromophenyl methyl sulphoxide 98%
Irritant
4-Bromophenyl methyl sulphoxide 98%
Catalog # APOSOR3341-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Bromophenyl methyl sulphoxide 98%
Catalog # APOSOR3341-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :219.099 g/mol
Melting Pt:78…80 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:934-71-4
Formula:C₇H₇BrOS
MDL Number:MFCD00159074