Order Entry
Switzerland
ContactUsLinkComponent
(3Z)-4-(Dimethylamino)-3-phenylbut-3-en-2-one, tech
(3Z)-4-(Dimethylamino)-3-phenylbut-3-en-2-one, tech
Catalog # APOSOR33355-1G
Supplier:  Apollo Scientific
CAS Number:  
(3Z)-4-(Dimethylamino)-3-phenylbut-3-en-2-one, tech
Catalog # APOSOR33355-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :189.254 g/mol
Storage Temperature:Ambient
CAS Number:343569-91-5
Formula:C₁₂H₁₅NO
MDL Number:MFCD00975170