Order Entry
Switzerland
ContactUsLinkComponent
7-Chloro-6-(2-chloroethyl)-2,5-dimethylpyrazolo[1,5-a]pyrimidine, tech
7-Chloro-6-(2-chloroethyl)-2,5-dimethylpyrazolo[1,5-a]pyrimidine, tech
Catalog # APOSOR33287-1G
Supplier:  Apollo Scientific
CAS Number:  
7-Chloro-6-(2-chloroethyl)-2,5-dimethylpyrazolo[1,5-a]pyrimidine, tech
Catalog # APOSOR33287-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :244.12 g/mol
Storage Temperature:Ambient
CAS Number:129909-71-3
Formula:C₁₀H₁₁Cl₂N₃
MDL Number:MFCD03305817