1-(2,5-Dichlorothiophen-3-yl)-3-(dimethylamino)propan-1-ol, tech
Catalog # APOSOR33014-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:254.177 g/mol
Storage Temperature:Ambient
CAS Number:439110-94-8
Formula:C₉H₁₃Cl₂NOS
MDL Number:MFCD02661707