Order Entry
Switzerland
ContactUsLinkComponent
1-[4-(1H-Pyrrol-1-yl)phenyl]ethan-1-ol, tech
1-[4-(1H-Pyrrol-1-yl)phenyl]ethan-1-ol, tech
Catalog # APOSOR32915-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[4-(1H-Pyrrol-1-yl)phenyl]ethan-1-ol, tech
Catalog # APOSOR32915-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :187.238 g/mol
Storage Temperature:Ambient
CAS Number:439110-83-5
Formula:C₁₂H₁₃NO
MDL Number:MFCD03012945