Order Entry
Switzerland
ContactUsLinkComponent
(2Z)-3-(Dimethylamino)-1-(4-methoxyphenyl)but-2-en-1-one, tech
(2Z)-3-(Dimethylamino)-1-(4-methoxyphenyl)but-2-en-1-one, tech
Catalog # APOSOR32559-1G
Supplier:  Apollo Scientific
CAS Number:  
(2Z)-3-(Dimethylamino)-1-(4-methoxyphenyl)but-2-en-1-one, tech
Catalog # APOSOR32559-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.28 g/mol
Storage Temperature:Ambient
CAS Number:1164513-83-0
Formula:C₁₃H₁₇NO₂
MDL Number:MFCD01814634