Order Entry
Switzerland
ContactUsLinkComponent
4-[(1E)-(Methoxyimino)methyl]-1-methyl-3-phenyl-1H-pyrazol-5-ol, tech
4-[(1E)-(Methoxyimino)methyl]-1-methyl-3-phenyl-1H-pyrazol-5-ol, tech
Catalog # APOSOR32437-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[(1E)-(Methoxyimino)methyl]-1-methyl-3-phenyl-1H-pyrazol-5-ol, tech
Catalog # APOSOR32437-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :231.251 g/mol
Storage Temperature:Ambient
CAS Number:321533-56-6
Formula:C₁₂H₁₃N₃O₂
MDL Number:MFCD01315295