1-(3-(Piperidin-4-yl)phenyl)ethan-1-ol hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2256054-71-2
:
Formula:C₁₃H₁₉NO·ClH
MDL Number:MFCD30099235
:241.757 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together