
2-(4-Chlorophenyl)-4'-methylacetophenone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:62006-19-3
:
Formula:C₁₅H₁₃ClO
MDL Number:MFCD11210879
:244.716 g/mol
Melting Pt:112…114 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together