
2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-ol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:651030-55-6
:
Formula:C₁₄H₂₁BO₃
MDL Number:MFCD18733921
:248.126 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together