4-Bromo-6-methoxy-2-propylquinoline hydrobromide
Catalog # APOSOR309308-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:361.072 g/mol
Storage Temperature:Ambient
CAS Number:1204811-54-0
Formula:C₁₃H₁₄BrNO·BrH
MDL Number:MFCD12674909