Order Entry
Switzerland
ContactUsLinkComponent
[2-[2-(2-Hydroxy-ethoxy)-ethoxy]-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester
Irritant
[2-[2-(2-Hydroxy-ethoxy)-ethoxy]-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester
Catalog # APOSOR307804.1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
[2-[2-(2-Hydroxy-ethoxy)-ethoxy]-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester
Catalog # APOSOR307804.1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :371.427 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:560088-66-6
Formula:C₂₁H₂₅NO₅
MDL Number:MFCD08669697