Order Entry
Switzerland
ContactUsLinkComponent
1-(3-Chlorophenyl)cyclopropanecarbonitrile
1-(3-Chlorophenyl)cyclopropanecarbonitrile
Catalog # APOSOR307751-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3-Chlorophenyl)cyclopropanecarbonitrile
Catalog # APOSOR307751-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.63 g/mol
Storage Temperature:Ambient
CAS Number:124276-32-0
Formula:C₁₀H₈ClN
MDL Number:MFCD01314305