3-Amino-3-(4-isopropyl-phenyl)-propan-1-ol
Catalog # APOSOR307626-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:193.285 g/mol
Storage Temperature:Ambient
CAS Number:955287-40-8
Formula:C₁₂H₁₉NO
MDL Number:MFCD07366613