Order Entry
Switzerland
ContactUsLinkComponent
2-(Aminomethyl)-1H-imidazole-5-carboxylic acid, 2-BOC protected
Irritant
2-(Aminomethyl)-1H-imidazole-5-carboxylic acid, 2-BOC protected
Katalognr. APOSOR30644-50MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(Aminomethyl)-1H-imidazole-5-carboxylic acid, 2-BOC protected
Katalognr. APOSOR30644-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :241.244 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:867340-37-2
Formula:C₁₀H₁₅N₃O₄
MDL Number:MFCD10703328