

(Diphenylphosphoryl)methanol
Catalog # APOSOR25804-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:232.215 g/mol
:GHS07
:GHS08
Storage Temperature:Refrigerator
CAS Number:884-74-2
Formula:C₁₃H₁₃O₂P
MDL Number:MFCD00087496