(4,5-Dichloro-1H-imidazol-1-yl)methyl (4-chlorophenyl)carbamate
Catalog # APOSOR25372-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:320.559 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:650592-36-2
Formula:C₁₁H₈Cl₃N₃O₂
MDL Number:MFCD00114451