
Ethyl 4-(iodomethyl)piperidine-4-carboxylate, N-BOC protected
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:213013-98-0
:
Formula:C₁₄H₂₄INO₄
MDL Number:MFCD09952119
:397.249 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together