3-(2,5-Dichloro-thien-3-yl)-1H-pyrazole
Catalog # APOSOR183461-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.091 g/mol
Storage Temperature:Ambient
CAS Number:166196-62-9
Formula:C₇H₄Cl₂N₂S
MDL Number:MFCD00112160