3-Methyl-1H-indole, N-BOC protected
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: tert-Butyl 3-methyl-1H-indole-1-carboxylate
CAS Number:89378-43-8
:
Formula:C₁₄H₁₇NO₂
MDL Number:MFCD13563142
:231.29 g/mol
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together