2,5-Dichloro-4-methoxyaniline
Catalog # APOSOR17402-10G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:192.043 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:101251-23-4
Formula:C₇H₇Cl₂NO
MDL Number:MFCD16606083