3-Bromo-2-methyl-1H-indole, N-BOC protected 98%
Catalog # APOSOR1732-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3-Bromo-1-(tert-butoxycarbonyl)-2-methyl-1H-indole, tert-Butyl 3-bromo-2-methyl-1H-indole-1-carboxylate, 1-Boc-3-Bromo-2-methylindole
:
:310.186 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:775305-12-9
Formula:C₁₄H₁₆BrNO₂
MDL Number:MFCD05864780