4-(Prop-1-ylamino)piperidine, N1-BOC protected 97%
Catalog # APOSOR16031-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Packung:Bottle
- VE:5 g
- Seq:3173
Specifications
About this item
:
:242.358 g/mol
Storage Temperature:Raumtemperatur
CAS Number:301225-58-1
Formula:C₁₃H₂₆N₂O₂
MDL Number:MFCD07367771