Order Entry
Switzerland
ContactUsLinkComponent
1-(Aminomethyl)-3-azabicyclo[3.1.0]hexane, 1-BOC protected
Irritant
1-(Aminomethyl)-3-azabicyclo[3.1.0]hexane, 1-BOC protected
Catalog # APOSOR16003-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(Aminomethyl)-3-azabicyclo[3.1.0]hexane, 1-BOC protected
Catalog # APOSOR16003-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :212.289 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:134574-96-2
Formula:C₁₁H₂₀N₂O₂
MDL Number:MFCD11035741