Order Entry
Switzerland
ContactUsLinkComponent
[2-(4-Bromophenoxy)phenyl]methanol
[2-(4-Bromophenoxy)phenyl]methanol
Catalog # APOSOR110491-1G
Supplier:  Apollo Scientific
CAS Number:  
[2-(4-Bromophenoxy)phenyl]methanol
Catalog # APOSOR110491-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :279.129 g/mol
Storage Temperature:Ambient
CAS Number:74744-79-9
Formula:C₁₃H₁₁BrO₂
MDL Number:MFCD11190400