SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
3-Iodo-1H-indole-2-carboxylic acid
3-Iodo-1H-indole-2-carboxylic acid
3-Iodo-1H-indole-2-carboxylic acid
APOSOR110213-1G
:
Apollo Scientific
:
3-Iodo-1H-indole-2-carboxylic acid
APOSOR110213-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
287.054 g/mol
:
GHS07
:
Ambient
:
167631-58-5
:
C₉H₆INO₂
:
MFCD06657947