
Diethyl (3-bromoprop-1-(E)-en-1-yl)phosphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:66498-59-7
:
Formula:C₇H₁₄BrO₃P
MDL Number:MFCD00800262
:257.062 g/mol
Boiling Pt:75 °C
Density:1.34 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together