1,5-Anhydro-D-mannitol ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:492-93-3
:
Formula:C₆H₁₂O₅
MDL Number:MFCD00067387
:164.156 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together