AccuScript high-fidelity reverse transcriptase
About this item
AccuScript High-Fidelity Reverse Transcriptase delivers the highest RT accuracy currently available.
- MMLV-RT derivative combined with 3 to 5’ exonuclease reduces errors
- High yields of full length cDNA up to 20 kb;RT-PCR kit synthesises cDNA from 10 to 1000 ng of total RNA or 0,1 to 10 ng of poly(A)+ RNA
- Stringent quality control procedures ensure high yields of premium cDNA
AccuScript High-Fidelity Reverse Transcriptase (RT) delivers 3,7 times more accuracy than other commercially available RTs and 6,6 times more accuracy than AMV RT. The AccuScript PfuUltra II RT-PCR kit provides a complete system in applications that demand high fidelity, specificity, and throughput.
Caution: For research use only. Not for use in diagnostic procedures.
2 Options Available Below
Product Details & Documents
AccuScript High-Fidelity Reverse Transcriptase delivers the highest RT accuracy currently available.
- MMLV-RT derivative combined with 3 to 5’ exonuclease reduces errors
- High yields of full length cDNA up to 20 kb;RT-PCR kit synthesises cDNA from 10 to 1000 ng of total RNA or 0,1 to 10 ng of poly(A)+ RNA
- Stringent quality control procedures ensure high yields of premium cDNA
AccuScript High-Fidelity Reverse Transcriptase (RT) delivers 3,7 times more accuracy than other commercially available RTs and 6,6 times more accuracy than AMV RT. The AccuScript PfuUltra II RT-PCR kit provides a complete system in applications that demand high fidelity, specificity, and throughput.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Product Information
- DetailsNo. of reactionsPkAvailabilityPrice
- AGLS600089AccuScript reverse transcriptaseAccuScript reverse transcriptaseEngineered to deliver the highest transcription accuracy while promoting full-length cDNA synthesis, yield, and RT-PCR performance50 reactions1Log in for availability1 * 1 ST
- High fidelity reverse transcriptase for long target lengths. Ideal for low RNA input and high specificity. Shorter PCR extension times with PfuUltra II with high accuracy. Kit includes dNTP, dTT, oligodT and random primers. Capable of amplifying cDNA of 0,1 - 9,6 kb in length ;10 - 1000 ng of total RNA or 0,1 - 10 ng of poly(A)+ RNA input50 reactionsLog in for availability1 * 1 ST