

Salzsäure 36,5 - 38,0% NF, Multikompendial, erfüllt die analytischen Spezifikationen von BP, Ph. Eur., J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
ADR:8,II
:Management of Change (MOC) category = R
:36.46 g/mol
Storage Temperature:Raumtemperatur
EINECS:231-595-7
:GHS05
:GHS07
Specification Test Results
- NF - Identification:Passes Test
- NF - Residue on Ignition:≤ 0.008 %
- NF - Bromide or Iodide:Passes Test
- NF - Free Bromine or Chlorine:Passes Test
- NF - Sulfate (SO₄):Passes Test
- NF - Sulfite (SO₃):Passes Test
- NF - Assay (HCl):36.5 - 38.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Free Chlorine (as Cl₂):≤ 4 ppm
- EP/BP - Sulfate (SO₄):≤ 20 ppm
- EP/BP - Residue on Evaporation:≤ 0.01 %
- EP/BP - Assay (HCl):35.0 - 39.0 %
- JP - Assay (HCl):35.0 - 38.0 %
- JP - Identification:Passes Test
- JP - Residue on ignition, mg/10mL:≤ 1.0
- JP - Bromide or Iodide:Passes Test
- JP - Bromine or Chlorine:Passes Test
- JP - Sulfate (SO₄):Passes Test
- JP - Sulfite (SO₃):Passes Test
- JP - Arsenic (As):≤ 1 ppm
- JP - Heavy Metals (as Pb):≤ 5 ppm
- JP - Mercury (Hg):≤ 0.04 ppm
- ChP - Assay:36.0 - 38.0 %
- ChP - Identification:Passes Test
- ChP - Arsenic (As):≤ 0.0001 %
- ChP - Bromide or Iodide:Passes Test
- ChP - Free Chlorine or Bromine:Passes Test
- ChP - Heavy Metals:≤ 0.0002 %
- ChP - Iron (Fe):≤ 0.0001 %
- ChP - Residue on Ignition:≤ 0.002 %
- ChP - Sulfate:≤ 0.0005 %
- ChP - Sulfite:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_BAKRCH09
SDS
spec_BAKRCH09-03 spec_BAKRCH09-07
Product specification
Frequently Bought Together