
2-Methyl-5-isopropylaniline (synthetic) ≥94% (by GC)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2051-53-8
:
Formula:C₁₀H₁₅N
MDL Number:MFCD00044503
Density:0.95 g/cm³
:149.24 g/mol
:GHS07
Specification Test Results
- Appearance:Clear colorless to yellow to brown liquid
- Infrared spectrum:Conforms
- Refractive index:1.5350 to 1.5450 (20°C, 589 nm)
- Water:≤2.5 %
- GC:≥94 %
Specification Test Results
Frequently Bought Together