Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Propionic acid ≥99.7%, GPR RECTAPUR®

Corrosive
Flammable
Irritant
Supplier:   VWR Chemicals
Propionic acid ≥99.7%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Propionic acid ≥99.7%, GPR RECTAPUR®

6 Options Available Below

Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Boiling Pt:140…142 °C
  • Flash Pt:54 °C
  • Storage Temperature:Ambient
  • UN:3463
  • ADR:8,II
Specifications for Product Family

Specification Test Results

  • Assay (GC) :
    min 99.0%
  • Identity (IR) :
    passes test
  • Density (d 20°C/4°C):
    0,993 – 0,994
  • Water (KF):
    max 0.30%
  • Appearance:
    clear liquid
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.