Order Entry
Switzerland
ContactUsLinkComponent
Potassium permanganate solution 0.002 mol/l (0.01 N) (20°C), Titripur®, Supelco®
Potassium permanganate solution 0.002 mol/l (0.01 N) (20°C), Titripur®, Supelco®
Catalog # 1.60322.1003
Supplier:  Merck
CAS Number:  
Potassium permanganate solution 0.002 mol/l (0.01 N) (20°C), Titripur®, Supelco®
Catalog # 1.60322.1003
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Bottle with RFID tag
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :158.03 g/mol
Storage Temperature:Ambient
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364