Order Entry
Switzerland
ContactUsLinkComponent
Propionaldehyde 2,4-dinitrophenylhydrazone analytical standard, for environmental analysis, Sigma-Aldrich®
Propionaldehyde 2,4-dinitrophenylhydrazone analytical standard, for environmental analysis, Sigma-Aldrich®
Catalog # 429546-20MG
CAS Number:  
undefined
Propionaldehyde 2,4-dinitrophenylhydrazone analytical standard, for environmental analysis, Sigma-Aldrich®
Catalog # 429546-20MG
CAS Number:  

Specifications

  • Pk:
    20 mg
  • Seq:
    2305

Specifications

About this item

 :
Melting Pt:152…155 °C (lit.)
 :238.20 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:725-00-8
Formula:C₉H₁₀N₄O₄
MDL Number:MFCD00191484