Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,3'-Dioctadecyloxacarbocyanine perchlorate (DiO)
DiO, Sigma-Aldrich®
DiO, Sigma-Aldrich®
DiO, Sigma-Aldrich®
D4292-20MG
:
SIGMA ALDRICH MICROSCOPY
:
DiO, Sigma-Aldrich®
D4292-20MG
:
SIGMA ALDRICH MICROSCOPY
:
Pk:
20 mg
Seq:
2305
:
:
881.721 g/mol
:
GHS07
:
Refrigerator
:
34215-57-1
:
C₅₃H₈₅ClN₂O₆