Order Entry
Switzerland
ContactUsLinkComponent
Di-8-ANEPPS ≥95% (by HPLC), Sigma-Aldrich®
Di-8-ANEPPS ≥95% (by HPLC), Sigma-Aldrich®
Catalog # D1065-5MG
CAS Number:  
undefined
Di-8-ANEPPS ≥95% (by HPLC), Sigma-Aldrich®
Catalog # D1065-5MG
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :592.87 g/mol
Storage Temperature:Refrigerator
CAS Number:157134-53-7
Formula:C₃₆H₅₂N₂O₃S
MDL Number:MFCD03453608