
Demecolcine ≥98% (by HPLC), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:477-30-5
:
Formula:C₂₁H₂₅NO₅
MDL Number:MFCD00075459
Melting Pt:184 °C
Storage Temperature:Refrigerator
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:371.433 g/mol
EINECS:207-514-6
UN:2811
:GHS06
Specification Test Results
- Appearance (colour):Faint yellow to yellow and faint beige
- Appearance (form):Powder
- Purity (hplc area %):≥98%
- Solubility (colour):Faint yellow to very dark yellow and faint green-yellow to very dark green-yellow
- Solubility (turbidity):Clear
- Solubility (method):10 mg plus 1 ml of chloroform
- Infrared spectrum:Conforms to structure
Specification Test Results
Frequently Bought Together