


Fast Blue BB base 15 mg/ml in Salzsäure 4M stabilisiert, Sigma-Aldrich®
Catalog # 917-10ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- VE:10 mL
- Seq:2275
Specifications
About this item
:
:300.36 g/mol
ADR:8,II
:GHS08
:GHS05
:GHS07
Storage Temperature:Kühlschrank
:1
CAS Number:120-00-3
Formula:C₁₇H₂₀N₂O₃
MDL Number:MFCD00009091