Order Entry
Switzerland
ContactUsLinkComponent
Trisodium citrate solution, Sigma-Aldrich®
Trisodium citrate solution, Sigma-Aldrich®
Catalog # 854-200ML
CAS Number:  
Trisodium citrate solution, Sigma-Aldrich®
Catalog # 854-200ML
CAS Number:  

Specifications

  • Pk:
    200 mL
  • Seq:
    2530

Specifications

About this item

 :
 :258.07 g/mol
Storage Temperature:Refrigerator
CAS Number:68-04-2
Formula:C₆H₅Na₃O₇
MDL Number:MFCD00012462